Product Details

SNP ID
rs146550775
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56232147 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATATACTTTATTGATAACCTGAT[C/T]TTTCCCAAACATTTTTTTCATTGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5T2 PubMed Links

Gene Details

Gene
OR5T2
Gene Name
olfactory receptor family 5 subfamily T member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004746.1 1039 Missense Mutation AAT,GAT N347D NP_001004746.1

View Full Product Details