Product Details

SNP ID
rs147892497
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:46720809 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAGCTTGTGAGACAGCGAGGAC[A/G]CCTCGAGATAAGCTTGCTGCATGTC
Phenotype
MIM: 176930
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
F2 PubMed Links

Gene Details

Gene
F2
Gene Name
coagulation factor II, thrombin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000506.4 307 Silent Mutation ACA,ACG T95T NP_000497.1
NM_001311257.1 307 Silent Mutation ACA,ACG T79T NP_001298186.1

View Full Product Details