Product Details

SNP ID
rs147991689
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59713447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGGACCAGCTCATGATGAGCCT[C/G]TATCGCAGAGGGTGACAGATCGCTA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR10V1 PubMed Links

Gene Details

Gene
OR10V1
Gene Name
olfactory receptor family 10 subfamily V member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005324.1 399 Nonsense Mutation TAC,TAG Y133* NP_001005324.1

View Full Product Details