Product Details

SNP ID
rs148579088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:93743777 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCACTCAAATGGTAACCAGTTT[A/G]GTTGCGATAGTTGGTCTACCCCTCC
Phenotype
MIM: 615810 MIM: 612823
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf54 PubMed Links

Gene Details

Gene
C11orf54
Gene Name
chromosome 11 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286067.1 888 Intron NP_001272996.1
NM_001286068.1 888 Intron NP_001272997.1
NM_001286069.1 888 Intron NP_001272998.1
NM_001286070.1 888 Intron NP_001272999.1
NM_001286071.1 888 Intron NP_001273000.1
NM_014039.3 888 Intron NP_054758.2
XM_006718824.1 888 Intron XP_006718887.1
XM_011542781.2 888 Intron XP_011541083.1
XM_011542782.2 888 Intron XP_011541084.1
XM_017017613.1 888 UTR 5 XP_016873102.1
XM_017017614.1 888 Intron XP_016873103.1
XM_017017615.1 888 Intron XP_016873104.1
XM_017017616.1 888 Intron XP_016873105.1
XM_017017617.1 888 Intron XP_016873106.1
XM_017017618.1 888 Intron XP_016873107.1
XM_017017619.1 888 Intron XP_016873108.1
Gene
SNORA40
Gene Name
small nucleolar RNA, H/ACA box 40
There are no transcripts associated with this gene.

Gene
TAF1D
Gene Name
TATA-box binding protein associated factor, RNA polymerase I subunit D
There are no transcripts associated with this gene.

View Full Product Details