Product Details

SNP ID
rs148657140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:27496329 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGGGTGATAATTTATATTTGTA[A/G]TTTACAAATCAAACATCTATCATAC
Phenotype
MIM: 612332
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LIN7C PubMed Links

Gene Details

Gene
LIN7C
Gene Name
lin-7 homolog C, crumbs cell polarity complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018362.3 2941 UTR 3 NP_060832.1

View Full Product Details