Product Details

SNP ID
rs149175291
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:117428708 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCGAGCCAGGTTCCGGATGGAGG[C/T]CTCACGGGGTGGGACCACGGGGCAG
Phenotype
MIM: 611782
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DSCAML1 PubMed Links

Gene Details

Gene
DSCAML1
Gene Name
DS cell adhesion molecule like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020693.3 4241 Missense Mutation ACC,GCC T1988A NP_065744.2
XM_011542917.1 4241 Missense Mutation ACC,GCC T1940A XP_011541219.1
XM_011542918.1 4241 Missense Mutation ACC,GCC T1852A XP_011541220.1
XM_011542919.2 4241 Missense Mutation ACC,GCC T1501A XP_011541221.1
XM_011542920.2 4241 Missense Mutation ACC,GCC T1418A XP_011541222.1
XM_011542921.2 4241 Missense Mutation ACC,GCC T1219A XP_011541223.1
XM_011542924.2 4241 Missense Mutation ACC,GCC T1145A XP_011541226.1
XM_011542925.2 4241 Missense Mutation ACC,GCC T1139A XP_011541227.1

View Full Product Details