Product Details

SNP ID
rs149817552
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59829303 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATGAAAATTTTATTTTCATGAG[C/T]CATAGATGTTGAGACTCCAGTGAAC
Phenotype
MIM: 609342
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GIF PubMed Links

Gene Details

Gene
GIF
Gene Name
gastric intrinsic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005142.2 1469 UTR 3 NP_005133.2
XM_011544939.2 1469 UTR 3 XP_011543241.1

View Full Product Details