Product Details
- SNP ID
-
rs149828734
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:8696649 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGATAAACACCTCCGACACCAGCC[C/T]ATTGAGGGTATTACATTCTGGAAGG
- Phenotype
-
MIM: 603637
MIM: 140750
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RPL27A
PubMed Links
Gene Details
- Gene
- RPL27A
- Gene Name
- ribosomal protein L27a
There are no transcripts associated with this gene.
- Gene
- ST5
- Gene Name
- suppression of tumorigenicity 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005418.3 |
3464 |
Missense Mutation |
AGG,GGG |
R1024G |
NP_005409.3 |
NM_139157.2 |
3464 |
Missense Mutation |
AGG,GGG |
R604G |
NP_631896.1 |
NM_213618.1 |
3464 |
Missense Mutation |
AGG,GGG |
R1024G |
NP_998783.1 |
XM_005253077.1 |
3464 |
Missense Mutation |
AGG,GGG |
R1054G |
XP_005253134.1 |
XM_005253083.2 |
3464 |
Missense Mutation |
AGG,GGG |
R604G |
XP_005253140.1 |
XM_011520309.1 |
3464 |
Missense Mutation |
AGG,GGG |
R1074G |
XP_011518611.1 |
XM_011520310.1 |
3464 |
Missense Mutation |
AGG,GGG |
R1064G |
XP_011518612.1 |
XM_011520311.2 |
3464 |
Missense Mutation |
AGG,GGG |
R1044G |
XP_011518613.1 |
XM_011520312.1 |
3464 |
Missense Mutation |
AGG,GGG |
R1044G |
XP_011518614.1 |
XM_011520313.1 |
3464 |
Missense Mutation |
AGG,GGG |
R1044G |
XP_011518615.1 |
XM_011520314.1 |
3464 |
Missense Mutation |
AGG,GGG |
R1044G |
XP_011518616.1 |
XM_011520315.1 |
3464 |
Missense Mutation |
AGG,GGG |
R1044G |
XP_011518617.1 |
XM_011520316.1 |
3464 |
Missense Mutation |
AGG,GGG |
R1044G |
XP_011518618.1 |
XM_011520317.1 |
3464 |
Missense Mutation |
AGG,GGG |
R1044G |
XP_011518619.1 |
XM_011520318.1 |
3464 |
Missense Mutation |
AGG,GGG |
R1024G |
XP_011518620.1 |
XM_011520319.2 |
3464 |
Missense Mutation |
AGG,GGG |
R864G |
XP_011518621.1 |
XM_011520320.1 |
3464 |
Intron |
|
|
XP_011518622.1 |
XM_011520321.2 |
3464 |
Intron |
|
|
XP_011518623.1 |
XM_011520322.1 |
3464 |
Intron |
|
|
XP_011518624.1 |
XM_011520323.2 |
3464 |
Missense Mutation |
AGG,GGG |
R654G |
XP_011518625.2 |
XM_011520324.2 |
3464 |
Missense Mutation |
AGG,GGG |
R634G |
XP_011518626.2 |
XM_011520325.1 |
3464 |
Missense Mutation |
AGG,GGG |
R624G |
XP_011518627.1 |
XM_011520326.1 |
3464 |
Missense Mutation |
AGG,GGG |
R624G |
XP_011518628.1 |
XM_011520327.1 |
3464 |
Missense Mutation |
AGG,GGG |
R624G |
XP_011518629.1 |
XM_011520328.1 |
3464 |
Missense Mutation |
AGG,GGG |
R624G |
XP_011518630.1 |
XM_011520329.1 |
3464 |
Missense Mutation |
AGG,GGG |
R604G |
XP_011518631.1 |
XM_017018182.1 |
3464 |
Missense Mutation |
AGG,GGG |
R1044G |
XP_016873671.1 |
XM_017018183.1 |
3464 |
Missense Mutation |
AGG,GGG |
R1044G |
XP_016873672.1 |
XM_017018184.1 |
3464 |
Missense Mutation |
AGG,GGG |
R864G |
XP_016873673.1 |
XM_017018185.1 |
3464 |
Missense Mutation |
AGG,GGG |
R864G |
XP_016873674.1 |
XM_017018186.1 |
3464 |
Missense Mutation |
AGG,GGG |
R844G |
XP_016873675.1 |
XM_017018187.1 |
3464 |
Missense Mutation |
AGG,GGG |
R624G |
XP_016873676.1 |
View Full Product Details