Product Details

SNP ID
rs150516929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:111908832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATGGGAATGGTGCGCTCAGGGC[C/T]AGAGACCTGTTTCCTTGGTCCATTC
Phenotype
MIM: 123590 MIM: 602179
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRYAB PubMed Links

Gene Details

Gene
CRYAB
Gene Name
crystallin alpha B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289807.1 767 Missense Mutation AGC,GGC S154G NP_001276736.1
NM_001289808.1 767 Missense Mutation AGC,GGC S154G NP_001276737.1
NM_001885.2 767 Missense Mutation AGC,GGC S154G NP_001876.1
XM_011542608.1 767 Missense Mutation AGC,GGC S154G XP_011540910.1
XM_011542609.2 767 Missense Mutation AGC,GGC S87G XP_011540911.1
Gene
HSPB2
Gene Name
heat shock protein family B (small) member 2
There are no transcripts associated with this gene.

Gene
HSPB2-C11orf52
Gene Name
HSPB2-C11orf52 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details