Product Details

SNP ID
rs150926439
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59835971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTTAGATGCAGAGGTGGGGCCAG[A/G]GCCAGGGTTGCTGGGTAGAGTTGGT
Phenotype
MIM: 609342
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GIF PubMed Links

Gene Details

Gene
GIF
Gene Name
gastric intrinsic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005142.2 944 Missense Mutation CCT,TCT P304S NP_005133.2
XM_011544939.2 944 Missense Mutation CCT,TCT P290S XP_011543241.1

View Full Product Details