Product Details
- SNP ID
-
rs138511907
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:12330119 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCGAGAGGATTCTACCCAGGAGGA[C/T]GAGGCCTATCACCAGGAACAGGACA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MANSC1
PubMed Links
Gene Details
- Gene
- MANSC1
- Gene Name
- MANSC domain containing 1
There are no transcripts associated with this gene.
View Full Product Details