Product Details

SNP ID
rs139512043
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49325597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGCTGTCACCTGGGGCGCCAGC[C/T]TAGTCTGGCTAAAAGAGTACTGGTT
Phenotype
MIM: 615229 MIM: 603872
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
C1QL4 PubMed Links

Gene Details

Gene
C1QL4
Gene Name
complement C1q like 4
There are no transcripts associated with this gene.

Gene
LOC101927267
Gene Name
uncharacterized LOC101927267
There are no transcripts associated with this gene.

Gene
TROAP
Gene Name
trophinin associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100620.1 601 Intron NP_001094090.1
NM_001278324.1 601 Intron NP_001265253.1
NM_005480.3 601 Missense Mutation CCT,CTT P145L NP_005471.3
XM_006719181.1 601 Missense Mutation CCT,CTT P145L XP_006719244.1
XM_011537723.1 601 Missense Mutation CCT,CTT P145L XP_011536025.1
XM_011537724.1 601 UTR 5 XP_011536026.1
XM_011537725.2 601 Intron XP_011536027.1

View Full Product Details