Product Details

SNP ID
rs139549004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:98593987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTCCCCTAGAAAGATGTTCTC[A/G]TCCGTGGCGCACCTGGCGCGGGCGA
Phenotype
MIM: 600370
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC25A3 PubMed Links

Gene Details

Gene
SLC25A3
Gene Name
solute carrier family 25 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002635.3 129 Silent Mutation TCA,TCG S3S NP_002626.1
NM_005888.3 129 Silent Mutation TCA,TCG S3S NP_005879.1
NM_213611.2 129 Silent Mutation TCA,TCG S3S NP_998776.1
Gene
SNORA53
Gene Name
small nucleolar RNA, H/ACA box 53
There are no transcripts associated with this gene.

View Full Product Details