Product Details

SNP ID
rs141803388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:124324447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTCGCTTTTTAATTAGAACGAC[A/G]TGTGTAAATGACAGAATACCACCAC
Phenotype
MIM: 615927 MIM: 600848
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM101A PubMed Links

Gene Details

Gene
FAM101A
Gene Name
family with sequence similarity 101 member A
There are no transcripts associated with this gene.

Gene
NCOR2
Gene Name
nuclear receptor corepressor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077261.3 8653 UTR 3 NP_001070729.2
NM_001206654.1 8653 UTR 3 NP_001193583.1
NM_006312.5 8653 UTR 3 NP_006303.4
Gene
ZNF664-FAM101A
Gene Name
filamin-interacting protein FAM101A
There are no transcripts associated with this gene.

View Full Product Details