Product Details

SNP ID
rs145432917
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:93678849 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCAGAGGTATTGGTGGAGGGACT[C/G]GTTCTTCAGTACCTCTACCAGGAAG
Phenotype
MIM: 603454
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CRADD PubMed Links

Gene Details

Gene
CRADD
Gene Name
CASP2 and RIPK1 domain containing adaptor with death domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320099.1 206 Silent Mutation CTC,CTG L25L NP_001307028.1
NM_001320100.1 206 Silent Mutation CTC,CTG L25L NP_001307029.1
NM_001320101.1 206 Silent Mutation CTC,CTG L25L NP_001307030.1
NM_003805.4 206 Silent Mutation CTC,CTG L25L NP_003796.1
XM_017020142.1 206 Silent Mutation CTC,CTG L25L XP_016875631.1
XM_017020143.1 206 Silent Mutation CTC,CTG L25L XP_016875632.1
XM_017020144.1 206 Silent Mutation CTC,CTG L25L XP_016875633.1
XM_017020145.1 206 Silent Mutation CTC,CTG L25L XP_016875634.1

View Full Product Details