Product Details

SNP ID
rs145971728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:132139054 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGCCCGCAGCCATCCTGACCTC[C/T]ACACTCTGAAAGTGACAAGCCCTGA
Phenotype
MIM: 612819
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX51 PubMed Links

Gene Details

Gene
DDX51
Gene Name
DEAD-box helicase 51
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175066.3 2258 UTR 3 NP_778236.2
XM_011538256.1 2258 Intron XP_011536558.1
Gene
NOC4L
Gene Name
nucleolar complex associated 4 homolog
There are no transcripts associated with this gene.

View Full Product Details