Product Details

SNP ID
rs146366019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:92737422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGGAAACACATCACAAACTCCC[G/A]ACCGATCAATTAGACCTGAAAAAGC
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
PLEKHG7 PubMed Links

Gene Details

Gene
PLEKHG7
Gene Name
pleckstrin homology and RhoGEF domain containing G7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004330.2 134 UTR 5 NP_001004330.1

View Full Product Details