Product Details

SNP ID
rs147109469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:31677325 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAAAAGAAAAAGAAAAAGAATC[A/G]CATACTCTCCAGACTGAAAAAGGGC
Phenotype
MIM: 615256
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMN1 PubMed Links

Gene Details

Gene
AMN1
Gene Name
antagonist of mitotic exit network 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113402.1 Intron NP_001106873.1
NM_001278411.1 Intron NP_001265340.1
NM_001278412.1 Intron NP_001265341.1
XM_006719054.3 Intron XP_006719117.1
XM_011520604.2 Intron XP_011518906.1
XM_017018963.1 Intron XP_016874452.1
XM_017018964.1 Intron XP_016874453.1
XM_017018965.1 Intron XP_016874454.1
Gene
ETFBKMT
Gene Name
electron transfer flavoprotein beta subunit lysine methyltransferase
There are no transcripts associated with this gene.

View Full Product Details