Product Details

SNP ID
rs149959487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:4911514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCGTGGTGATCAACATCTCCGGG[A/C]TGCGCTTCGAGACGCAGCTCAAGAC
Phenotype
MIM: 176260
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KCNA1 PubMed Links

Gene Details

Gene
KCNA1
Gene Name
potassium voltage-gated channel subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000217.2 1241 Missense Mutation ATG,CTG M46L NP_000208.2

View Full Product Details