Product Details

SNP ID
rs150854777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52839355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCAACTCCTCCAGACATGACAG[C/T]GCTGCCTCCCACCGAGGCTGCCAAG
Phenotype
MIM: 611159 MIM: 611160
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT78 PubMed Links

Gene Details

Gene
KRT78
Gene Name
keratin 78
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300814.1 1003 Missense Mutation ACT,GCT T331A NP_001287743.1
NM_173352.3 1003 Missense Mutation ACT,GCT T441A NP_775487.2
XM_011538010.1 1003 Missense Mutation ACT,GCT T409A XP_011536312.1
Gene
KRT79
Gene Name
keratin 79
There are no transcripts associated with this gene.

View Full Product Details