Product Details

SNP ID
rs141089354
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:52024921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATTGGTCTGCATACCATGTGGAA[C/T]GAGCATTTTGGGATTGGTGAGTTTG
Phenotype
MIM: 613666 MIM: 608969
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALG11 PubMed Links

Gene Details

Gene
ALG11
Gene Name
ALG11, alpha-1,2-mannosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004127.2 231 Silent Mutation AAC,AAT N397N NP_001004127.2
Gene
UTP14C
Gene Name
UTP14, small subunit processome component homolog C (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021645.5 231 UTR 5 NP_067677.4

View Full Product Details