Product Details

SNP ID
rs150028098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:24322494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTCTATGTGTTTTCTAGGACTGC[T/C]CTAATAAAGTACACAAACTGGGAGT
Phenotype
MIM: 614285
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
C1QTNF9 PubMed Links

Gene Details

Gene
C1QTNF9
Gene Name
C1q and tumor necrosis factor related protein 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303137.1 Intron NP_001290066.1
NM_001303138.1 Intron NP_001290067.1
NM_178540.4 Intron NP_848635.2
Gene
LINC00566
Gene Name
long intergenic non-protein coding RNA 566
There are no transcripts associated with this gene.

Gene
NUS1P3
Gene Name
NUS1 dehydrodolichyl diphosphate synthase subunit pseudogene 3
There are no transcripts associated with this gene.

View Full Product Details