Product Details

SNP ID
rs142227946
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20583944 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCAGCACTGTCAATATCTTCTC[C/T]AATGAGGTAACACTATGGTATTGGC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LOC254028 PubMed Links

Gene Details

Gene
LOC254028
Gene Name
uncharacterized LOC254028
There are no transcripts associated with this gene.

Gene
RNASE11
Gene Name
ribonuclease A family member 11 (inactive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145250.3 715 Silent Mutation TTA,TTG L177L NP_660293.1
Gene
RNASE12
Gene Name
ribonuclease A family member 12 (inactive)
There are no transcripts associated with this gene.

View Full Product Details