Product Details

SNP ID
rs142397893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24505594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGACAGCAGGGGGCTTTGCATC[C/T]GACCGTCCATAGGATACGATGCCCT
Phenotype
MIM: 118938
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CMA1 PubMed Links

Gene Details

Gene
CMA1
Gene Name
chymase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308083.1 545 Silent Mutation TCA,TCG S111S NP_001295012.1
NM_001836.4 545 Silent Mutation TCA,TCG S222S NP_001827.1
Gene
LOC101927045
Gene Name
uncharacterized LOC101927045
There are no transcripts associated with this gene.

View Full Product Details