Product Details

SNP ID
rs143371520
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:60509692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGCTTGAAGCACACTACCAGGA[A/G]GCTGAGAAGCTGCGTGGAAGACCCC
Phenotype
MIM: 606326
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C14orf39 PubMed Links

Gene Details

Gene
C14orf39
Gene Name
chromosome 14 open reading frame 39
There are no transcripts associated with this gene.

Gene
SIX6
Gene Name
SIX homeobox 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007374.2 473 Silent Mutation GAA,GAG E98E NP_031400.2

View Full Product Details