Product Details

SNP ID
rs145338066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21042231 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCATCCTTAGGTGTCCACGTATT[A/T]GGGACATGTAAGTGGAGAGGCATGA
Phenotype
MIM: 605272 MIM: 612484 MIM: 616956
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NDRG2 PubMed Links

Gene Details

Gene
NDRG2
Gene Name
NDRG family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282211.1 6 Intron NP_001269140.1
NM_001282212.1 6 Intron NP_001269141.1
NM_001282213.1 6 Intron NP_001269142.1
NM_001282214.1 6 Intron NP_001269143.1
NM_001282215.1 6 Intron NP_001269144.1
NM_001282216.1 6 Intron NP_001269145.1
NM_001320329.1 6 Intron NP_001307258.1
NM_016250.2 6 Intron NP_057334.1
NM_201535.1 6 Intron NP_963293.1
NM_201536.1 6 Intron NP_963294.1
NM_201537.1 6 Intron NP_963831.1
NM_201538.1 6 Intron NP_963832.1
NM_201539.1 6 Intron NP_963833.1
NM_201540.1 6 Intron NP_963834.1
NM_201541.1 6 Intron NP_963835.1
XM_011536996.2 6 Intron XP_011535298.1
XM_011536997.1 6 Intron XP_011535299.1
XM_011536998.1 6 Intron XP_011535300.1
XM_011536999.1 6 Intron XP_011535301.1
XM_011537001.1 6 Intron XP_011535303.1
XM_011537002.1 6 Intron XP_011535304.1
XM_017021480.1 6 Intron XP_016876969.1
XM_017021481.1 6 Intron XP_016876970.1
XM_017021482.1 6 Intron XP_016876971.1
XM_017021483.1 6 Intron XP_016876972.1
XM_017021484.1 6 Intron XP_016876973.1
XM_017021485.1 6 Intron XP_016876974.1
XM_017021486.1 6 Intron XP_016876975.1
XM_017021487.1 6 Intron XP_016876976.1
XM_017021488.1 6 Intron XP_016876977.1
XM_017021489.1 6 Intron XP_016876978.1
XM_017021490.1 6 Intron XP_016876979.1
XM_017021491.1 6 Intron XP_016876980.1
XM_017021492.1 6 Intron XP_016876981.1
XM_017021493.1 6 Intron XP_016876982.1
XM_017021494.1 6 Intron XP_016876983.1
XM_017021495.1 6 Intron XP_016876984.1
XM_017021496.1 6 Intron XP_016876985.1
XM_017021497.1 6 Intron XP_016876986.1
Gene
RNASE13
Gene Name
ribonuclease A family member 13 (inactive)
There are no transcripts associated with this gene.

Gene
RNASE7
Gene Name
ribonuclease A family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032572.3 6 UTR 5 NP_115961.2
Gene
TPPP2
Gene Name
tubulin polymerization promoting protein family member 2
There are no transcripts associated with this gene.

View Full Product Details