Product Details

SNP ID
rs138973347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41388449 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAAAAATACAGGAATATGTTACC[C/T]TATCAAGCGGAAGCTCATGCTGAAC
Phenotype
MIM: 606934 MIM: 612818
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDUFAF1 PubMed Links

Gene Details

Gene
NDUFAF1
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016013.3 1161 Missense Mutation AAG,AGG K278R NP_057097.2
XM_006720555.2 1161 Missense Mutation AAG,AGG K278R XP_006720618.1
XM_011521658.1 1161 Missense Mutation AAG,AGG K278R XP_011519960.1
XM_011521659.2 1161 Missense Mutation AAG,AGG K278R XP_011519961.1
XM_017022293.1 1161 Intron XP_016877782.1
Gene
NUSAP1
Gene Name
nucleolar and spindle associated protein 1
There are no transcripts associated with this gene.

View Full Product Details