Product Details

SNP ID
rs139360469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41848401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACGTCTCCTCTTCACCCAGACA[G/A]GAATGCAGGGGGAGGCCAGGCAATG
Phenotype
MIM: 606088 MIM: 605916
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
JMJD7-PLA2G4B PubMed Links

Gene Details

Gene
JMJD7-PLA2G4B
Gene Name
JMJD7-PLA2G4B readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198588.1 11446 Intron NP_001185517.1
NM_005090.3 11446 Intron NP_005081.1
Gene
PLA2G4B
Gene Name
phospholipase A2 group IVB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114633.1 11446 Intron NP_001108105.1
Gene
SPTBN5
Gene Name
spectrin beta, non-erythrocytic 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016642.3 11446 UTR 3 NP_057726.4
XM_017022299.1 11446 UTR 3 XP_016877788.1
XM_017022300.1 11446 UTR 3 XP_016877789.1
XM_017022301.1 11446 UTR 3 XP_016877790.1
XM_017022302.1 11446 UTR 3 XP_016877791.1
XM_017022303.1 11446 Intron XP_016877792.1
XM_017022304.1 11446 Intron XP_016877793.1
XM_017022305.1 11446 Intron XP_016877794.1

View Full Product Details