Product Details

SNP ID
rs144560619
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:101176462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAATATTTTACCCTTTAAAGAGAA[G/A]GGTCAAGAAGAGAAAATGCCAAATC
Phenotype
MIM: 608183
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CHSY1 PubMed Links

Gene Details

Gene
CHSY1
Gene Name
chondroitin sulfate synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014918.4 3093 UTR 3 NP_055733.2
XM_006720435.3 3093 UTR 3 XP_006720498.1
XM_011521364.2 3093 UTR 3 XP_011519666.1
XM_017022011.1 3093 UTR 3 XP_016877500.1

View Full Product Details