Product Details

SNP ID
rs144864886
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:23566207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCTGGGGCCTCTGCAGGTGGAG[C/G]CCCTAGCACGGCTGCGCACATCGAG
Phenotype
MIM: 603856
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR4508 PubMed Links

Gene Details

Gene
MIR4508
Gene Name
microRNA 4508
There are no transcripts associated with this gene.

Gene
MKRN3
Gene Name
makorin ring finger protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005664.3 901 Missense Mutation GCC,GGC A142G NP_005655.1

View Full Product Details