Product Details

SNP ID
rs149346134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78264793 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGGAGACCCAGTACTATGACAT[C/T]CTGGGCGTGAAGCCCAGCGCGTCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJA4 PubMed Links

Gene Details

Gene
DNAJA4
Gene Name
DnaJ heat shock protein family (Hsp40) member A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130182.1 147 Silent Mutation ATC,ATT I10I NP_001123654.1
NM_001130183.1 147 Intron NP_001123655.1
NM_018602.3 147 Silent Mutation ATC,ATT I39I NP_061072.3
XM_005254535.3 147 Silent Mutation ATC,ATT I10I XP_005254592.1
XM_006720601.1 147 Silent Mutation ATC,ATT I10I XP_006720664.1

View Full Product Details