Product Details

SNP ID
rs150204440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74845047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGAAAGTGCAGCTCAGAAGGCAGG[C/T]GAGAGAAAGGCTGAGGGGGAGAGAA
Phenotype
MIM: 606912 MIM: 613472
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6882 PubMed Links

Gene Details

Gene
MIR6882
Gene Name
microRNA 6882
There are no transcripts associated with this gene.

Gene
SCAMP2
Gene Name
secretory carrier membrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320778.1 1279 UTR 3 NP_001307707.1
NM_005697.4 1279 UTR 3 NP_005688.2
XM_017021856.1 1279 UTR 3 XP_016877345.1
Gene
ULK3
Gene Name
unc-51 like kinase 3
There are no transcripts associated with this gene.

View Full Product Details