Product Details

SNP ID
rs139079671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:74622101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGGTACTTCAAAGAAAGTCAAA[A/T]CCTAAGCCTGCCCAGGCCCAAAGAC
Phenotype
MIM: 614151
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RFWD3 PubMed Links

Gene Details

Gene
RFWD3
Gene Name
ring finger and WD repeat domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018124.3 4203 UTR 3 NP_060594.3
XM_005256021.4 4203 UTR 3 XP_005256078.1
XM_005256022.4 4203 UTR 3 XP_005256079.1
XM_006721228.3 4203 UTR 3 XP_006721291.1
XM_011523191.2 4203 UTR 3 XP_011521493.1
XM_017023391.1 4203 UTR 3 XP_016878880.1
XM_017023392.1 4203 UTR 3 XP_016878881.1

View Full Product Details