Product Details

SNP ID
rs141062694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31038534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCCTGAAGGACACGCAGGTGACT[C/T]GACAGGCCTTAAATGAGATCTCGGC
Phenotype
MIM: 186591
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928762 PubMed Links

Gene Details

Gene
LOC101928762
Gene Name
uncharacterized LOC101928762
There are no transcripts associated with this gene.

Gene
STX4
Gene Name
syntaxin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272095.1 1040 Nonsense Mutation CGA,TGA R119* NP_001259024.1
NM_001272096.1 1040 Nonsense Mutation CGA,TGA R195* NP_001259025.1
NM_004604.4 1040 Nonsense Mutation CGA,TGA R197* NP_004595.2
XM_005255521.1 1040 Nonsense Mutation CGA,TGA R193* XP_005255578.1
XM_011545925.2 1040 Nonsense Mutation CGA,TGA R54* XP_011544227.1

View Full Product Details