Product Details

SNP ID
rs141298817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4461861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGGGGTTGAGTCTCAGGGTCAG[C/G]TCGATGTCACGGTCGGGTCTCAGGG
Phenotype
MIM: 608382
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNAJA3 PubMed Links

Gene Details

Gene
DNAJA3
Gene Name
DnaJ heat shock protein family (Hsp40) member A3
There are no transcripts associated with this gene.

Gene
NMRAL1
Gene Name
NmrA-like family domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305141.1 1132 Silent Mutation GAC,GAG D273E NP_001292070.1
NM_001305142.1 1132 Silent Mutation GAC,GAG D273E NP_001292071.1
NM_020677.4 1132 Silent Mutation GAC,GAG D273E NP_065728.1
XM_006720905.3 1132 Missense Mutation GAC,GAG D210E XP_006720968.1
XM_006720906.2 1132 Missense Mutation GAC,GAG D209E XP_006720969.1
XM_011522567.2 1132 Intron XP_011520869.1
XM_017023485.1 1132 Silent Mutation GAC,GAG D309E XP_016878974.1
XM_017023486.1 1132 Intron XP_016878975.1
XM_017023487.1 1132 Intron XP_016878976.1
XM_017023488.1 1132 Intron XP_016878977.1
XM_017023489.1 1132 Missense Mutation GAC,GAG D209E XP_016878978.1
XM_017023490.1 1132 Missense Mutation GAC,GAG D210E XP_016878979.1
XM_017023491.1 1132 UTR 3 XP_016878980.1
XM_017023492.1 1132 UTR 3 XP_016878981.1

View Full Product Details