Product Details
- SNP ID
-
rs141964049
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:27788493 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGTTCCTCCTGGCCTGGCTCCAC[C/T]GCCCTCTGATGCCTAATTTCCCACA
- Phenotype
-
MIM: 616650
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GSG1L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113342885] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GSG1L
- Gene Name
- GSG1 like
- Gene
- KIAA0556
- Gene Name
- KIAA0556
There are no transcripts associated with this gene.
View Full Product Details