Product Details

SNP ID
rs142312247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30570262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGCCGGGGGCTGTGCTCCTGTG[C/G]TCGGGTCCCAGGCAGCATTTTTGGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF688 PubMed Links

Gene Details

Gene
ZNF688
Gene Name
zinc finger protein 688
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024683.1 575 Missense Mutation ACC,AGC T148S NP_001019854.1
NM_145271.3 575 Missense Mutation ACC,AGC T162S NP_660314.1
XM_005255139.2 575 Missense Mutation ACC,AGC T85S XP_005255196.1
XM_005255140.2 575 Missense Mutation ACC,AGC T85S XP_005255197.1
Gene
ZNF785
Gene Name
zinc finger protein 785
There are no transcripts associated with this gene.

View Full Product Details