Product Details

SNP ID
rs143307181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:8533556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACCACTGAATATTTTTAAAAGGT[A/G]AATGGCATGATCTGGTCCAGAGCTT
Phenotype
MIM: 611579
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM114 PubMed Links

Gene Details

Gene
TMEM114
Gene Name
transmembrane protein 114
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146336.1 Intron NP_001139808.1
NM_001290095.1 Intron NP_001277024.1
NM_001290097.1 Intron NP_001277026.1
NM_001290098.1 Intron NP_001277027.1

View Full Product Details