Product Details
- SNP ID
-
rs143572732
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:4463744 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGTGTGCCTGCAAGTGCTCAGCCC[A/G]ATGTTCTGGCCGACGTATTTTTCTG
- Phenotype
-
MIM: 608382
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DNAJA3
PubMed Links
Gene Details
- Gene
- DNAJA3
- Gene Name
- DnaJ heat shock protein family (Hsp40) member A3
There are no transcripts associated with this gene.
- Gene
- NMRAL1
- Gene Name
- NmrA-like family domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001305141.1 |
949 |
Silent Mutation |
ATC,ATT |
I212I |
NP_001292070.1 |
NM_001305142.1 |
949 |
Silent Mutation |
ATC,ATT |
I212I |
NP_001292071.1 |
NM_020677.4 |
949 |
Silent Mutation |
ATC,ATT |
I212I |
NP_065728.1 |
XM_006720905.3 |
949 |
Silent Mutation |
ATC,ATT |
I149I |
XP_006720968.1 |
XM_006720906.2 |
949 |
Silent Mutation |
ATC,ATT |
I148I |
XP_006720969.1 |
XM_011522567.2 |
949 |
Silent Mutation |
ATC,ATT |
I212I |
XP_011520869.1 |
XM_017023485.1 |
949 |
Silent Mutation |
ATC,ATT |
I248I |
XP_016878974.1 |
XM_017023486.1 |
949 |
Silent Mutation |
ATC,ATT |
I248I |
XP_016878975.1 |
XM_017023487.1 |
949 |
Silent Mutation |
ATC,ATT |
I248I |
XP_016878976.1 |
XM_017023488.1 |
949 |
Silent Mutation |
ATC,ATT |
I212I |
XP_016878977.1 |
XM_017023489.1 |
949 |
Silent Mutation |
ATC,ATT |
I148I |
XP_016878978.1 |
XM_017023490.1 |
949 |
Silent Mutation |
ATC,ATT |
I149I |
XP_016878979.1 |
XM_017023491.1 |
949 |
Intron |
|
|
XP_016878980.1 |
XM_017023492.1 |
949 |
Intron |
|
|
XP_016878981.1 |
View Full Product Details