Product Details

SNP ID
rs145741242
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30376475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATGCCATGATGAAGGAAGCCAG[C/T]GGTCCCATCAACTTCACCGTCTTCC
Phenotype
MIM: 612897 MIM: 613620
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LOC107983970 PubMed Links

Gene Details

Gene
LOC107983970
Gene Name
uncharacterized LOC107983970
There are no transcripts associated with this gene.

Gene
MYLPF
Gene Name
myosin light chain, phosphorylatable, fast skeletal muscle
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324458.1 471 Silent Mutation AGC,AGT S75S NP_001311387.1
NM_001324459.1 471 Silent Mutation AGC,AGT S75S NP_001311388.1
NM_013292.4 471 Silent Mutation AGC,AGT S75S NP_037424.2
Gene
SEPT1
Gene Name
septin 1
There are no transcripts associated with this gene.

Gene
TBC1D10B
Gene Name
TBC1 domain family member 10B
There are no transcripts associated with this gene.

Gene
ZNF48
Gene Name
zinc finger protein 48
There are no transcripts associated with this gene.

View Full Product Details