Product Details
- SNP ID
-
rs147440681
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:27789291 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGCAGAAATGGATAATGGATGGAT[G/A]GATGGTTGATGGATAATGGGCAGAT
- Phenotype
-
MIM: 616650
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GSG1L
PubMed Links
Gene Details
- Gene
- GSG1L
- Gene Name
- GSG1 like
- Gene
- KIAA0556
- Gene Name
- KIAA0556
There are no transcripts associated with this gene.
View Full Product Details