Product Details

SNP ID
rs150493488
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:372106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGTGGCGGCAGCAGCAAGGCTG[C/T]GCTCCCGGCCAGCAGGATGTGCCAG
Phenotype
MIM: 604853
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL28 PubMed Links

Gene Details

Gene
MRPL28
Gene Name
mitochondrial ribosomal protein L28
There are no transcripts associated with this gene.

Gene
TMEM8A
Gene Name
transmembrane protein 8A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021259.2 2326 Missense Mutation ACA,GCA T733A NP_067082.2

View Full Product Details