Product Details

SNP ID
rs151000137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:47081909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAACAAAAGGAAAGAAAGAGCTTA[T/C]GTCCACATTTCCAAGGTCTTTACAA
Phenotype
MIM: 607974
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
NETO2 PubMed Links

Gene Details

Gene
NETO2
Gene Name
neuropilin and tolloid like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201477.1 3282 UTR 3 NP_001188406.1
NM_018092.4 3282 UTR 3 NP_060562.3
XM_006721289.1 3282 UTR 3 XP_006721352.1
XM_006721291.1 3282 UTR 3 XP_006721354.1
XM_006721292.1 3282 Intron XP_006721355.1
XM_017023740.1 3282 UTR 3 XP_016879229.1

View Full Product Details