Product Details

SNP ID
rs138579421
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:80202411 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCCGCGCACGGCACCATCCCCA[A/G]CTACTCCAGGTGAGCAGCTGCCTCG
Phenotype
MIM: 607211 MIM: 605270
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CARD14 PubMed Links

Gene Details

Gene
CARD14
Gene Name
caspase recruitment domain family member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257970.1 2457 Missense Mutation AAC,AGC N737S NP_001244899.1
NM_024110.4 2457 Missense Mutation AAC,AGC N737S NP_077015.2
NM_052819.2 2457 Intron NP_438170.1
XM_011525212.1 2457 Missense Mutation AAC,AGC N737S XP_011523514.1
XM_011525213.1 2457 Missense Mutation AAC,AGC N737S XP_011523515.1
XM_011525215.1 2457 Missense Mutation AAC,AGC N737S XP_011523517.1
XM_011525216.1 2457 Missense Mutation AAC,AGC N737S XP_011523518.1
XM_011525217.1 2457 Missense Mutation AAC,AGC N737S XP_011523519.1
XM_011525218.2 2457 Missense Mutation AAC,AGC N737S XP_011523520.1
XM_017025057.1 2457 Missense Mutation AAC,AGC N736S XP_016880546.1
XM_017025058.1 2457 Intron XP_016880547.1
Gene
LOC105371920
Gene Name
uncharacterized LOC105371920
There are no transcripts associated with this gene.

Gene
SGSH
Gene Name
N-sulfoglucosamine sulfohydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000199.3 2457 Intron NP_000190.1
XM_005257582.3 2457 Intron XP_005257639.1
XM_005257583.4 2457 Intron XP_005257640.1
XM_011525126.1 2457 Intron XP_011523428.1
XM_011525127.2 2457 Intron XP_011523429.1
XM_017024951.1 2457 Intron XP_016880440.1
XM_017024952.1 2457 Intron XP_016880441.1

View Full Product Details