Product Details

SNP ID
rs140948391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5105859 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCTTTTCTGGCATGAACTCTCC[A/G]ATGTCTAATGAGCTCTGAGCACCAT
Phenotype
MIM: 194480 MIM: 616463
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928000 PubMed Links

Gene Details

Gene
LOC101928000
Gene Name
uncharacterized LOC101928000
There are no transcripts associated with this gene.

Gene
ZFP3
Gene Name
ZFP3 zinc finger protein
There are no transcripts associated with this gene.

Gene
ZNF232
Gene Name
zinc finger protein 232
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320952.1 1418 Missense Mutation CGG,TGG R425W NP_001307881.1
NM_001320953.1 1418 Missense Mutation CGG,TGG R407W NP_001307882.1
NM_001320954.1 1418 Missense Mutation CGG,TGG R407W NP_001307883.1
NM_001320955.1 1418 Missense Mutation CGG,TGG R406W NP_001307884.1
NM_014519.3 1418 Missense Mutation CGG,TGG R434W NP_055334.2
XM_011524006.2 1418 Missense Mutation CGG,TGG R386W XP_011522308.1
XM_017025021.1 1418 Missense Mutation CGG,TGG R434W XP_016880510.1
XM_017025022.1 1418 Missense Mutation CGG,TGG R358W XP_016880511.1
XM_017025023.1 1418 Missense Mutation CGG,TGG R349W XP_016880512.1

View Full Product Details