Product Details

SNP ID
rs141122143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:80202373 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCATGCCCACCGCGTGAACTCTTA[A/C]ACCATGAAGGATACTGCCGCGCACG
Phenotype
MIM: 607211 MIM: 605270
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CARD14 PubMed Links

Gene Details

Gene
CARD14
Gene Name
caspase recruitment domain family member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257970.1 2419 Silent Mutation TAA,TAC *724Y NP_001244899.1
NM_024110.4 2419 Silent Mutation TAA,TAC *724Y NP_077015.2
NM_052819.2 2419 Intron NP_438170.1
XM_011525212.1 2419 Nonsense Mutation TAA,TAC *724Y XP_011523514.1
XM_011525213.1 2419 Nonsense Mutation TAA,TAC *724Y XP_011523515.1
XM_011525215.1 2419 Nonsense Mutation TAA,TAC *724Y XP_011523517.1
XM_011525216.1 2419 Nonsense Mutation TAA,TAC *724Y XP_011523518.1
XM_011525217.1 2419 Nonsense Mutation TAA,TAC *724Y XP_011523519.1
XM_011525218.2 2419 Nonsense Mutation TAA,TAC *724Y XP_011523520.1
XM_017025057.1 2419 Nonsense Mutation TAA,TAC *723Y XP_016880546.1
XM_017025058.1 2419 Intron XP_016880547.1
Gene
LOC105371920
Gene Name
uncharacterized LOC105371920
There are no transcripts associated with this gene.

Gene
SGSH
Gene Name
N-sulfoglucosamine sulfohydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000199.3 2419 Intron NP_000190.1
XM_005257582.3 2419 Intron XP_005257639.1
XM_005257583.4 2419 Intron XP_005257640.1
XM_011525126.1 2419 Intron XP_011523428.1
XM_011525127.2 2419 Intron XP_011523429.1
XM_017024951.1 2419 Intron XP_016880440.1
XM_017024952.1 2419 Intron XP_016880441.1

View Full Product Details