Product Details

SNP ID
rs141400019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58087180 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGACTGCGCCACGCAGGCCATGGA[C/G]AAGTACAATATAGAGAAGGACATTG
Phenotype
MIM: 608942
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DYNLL2 PubMed Links

Gene Details

Gene
DYNLL2
Gene Name
dynein light chain LC8-type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080677.2 364 Missense Mutation GAC,GAG D30E NP_542408.1
Gene
LOC101927666
Gene Name
uncharacterized LOC101927666
There are no transcripts associated with this gene.

View Full Product Details