Product Details

SNP ID
rs142788854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42569568 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGCAGGAGGAGGAGGTGTCCAC[A/G]TTACGCAAGGATGTCACCGCCCTAA
Phenotype
MIM: 609855 MIM: 616498 MIM: 602976 MIM: 608665
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COASY PubMed Links

Gene Details

Gene
COASY
Gene Name
Coenzyme A synthase
There are no transcripts associated with this gene.

Gene
FAM134C
Gene Name
family with sequence similarity 134 member C
There are no transcripts associated with this gene.

Gene
MLX
Gene Name
MLX, MAX dimerization protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_170607.2 503 Silent Mutation ACA,ACG T200T NP_733752.1
NM_198204.1 503 Silent Mutation ACA,ACG T146T NP_937847.1
NM_198205.1 503 Silent Mutation ACA,ACG T116T NP_937848.1
XM_017024991.1 503 Silent Mutation ACA,ACG T170T XP_016880480.1
Gene
PSMC3IP
Gene Name
PSMC3 interacting protein
There are no transcripts associated with this gene.

View Full Product Details