Product Details

SNP ID
rs145103209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38299440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACTGATAGAGAGAACTGAACGA[A/G]TGAAGAAGACTATGGCATCACAAGT
Phenotype
MIM: 611850
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL45 PubMed Links

Gene Details

Gene
MRPL45
Gene Name
mitochondrial ribosomal protein L45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278279.2 495 Missense Mutation ATG,GTG M112V NP_001265208.2
NM_032351.5 495 Missense Mutation ATG,GTG M112V NP_115727.5

View Full Product Details