Product Details

SNP ID
rs146036461
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:59962854 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTATATGCTGCATAACAAGTTTGA[C/T]GCTCAGAATCAAAATATATGGAAGA
Phenotype
MIM: 615172
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101927755 PubMed Links

Gene Details

Gene
LOC101927755
Gene Name
uncharacterized LOC101927755
There are no transcripts associated with this gene.

Gene
RNFT1
Gene Name
ring finger protein, transmembrane 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016125.3 Intron NP_057209.3
Gene
TBC1D3P1-DHX40P1
Gene Name
TBC1D3P1-DHX40P1 readthrough, transcribed pseudogene
There are no transcripts associated with this gene.

View Full Product Details